Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: LWC06, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14332801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14332801

  • / Each
 

Description & Specifications

Specifications

Antigen DC-SIGN/CD209
Conjugate Unlabeled
Gene ID (Entrez) 30835
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Mouse
Format Affinity Purified
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CIRE.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Affinity Purified
Clone LWC06
Isotype IgG2a
Host Species Rat
Quantity 0.1mg