Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: LWC06, FITC, Novus Biologicals™

$131.00 - $329.00

Specifications

Antigen DC-SIGN/CD209
Clone LWC06
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CIRE.
Gene ID (Entrez) 30835
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14396501

 
novus biologicals canada
NBP14396501MG
0.1mg Each for $329.00

NBP14396503

 
novus biologicals canada
NBP1439650025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen DC-SIGN/CD209
Clone LWC06
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CIRE.
Gene ID (Entrez) 30835
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry