Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: LWC06, Biotin, Novus Biologicals™

$131.00 - $329.00

Specifications

Antigen DC-SIGN/CD209
Clone LWC06
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CIRE.
Gene ID (Entrez) 30835
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14356601

 
novus biologicals canada
NBP14356601MG
0.1mg Each for $329.00

NBP14356603

 
novus biologicals canada
NBP1435660025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen DC-SIGN/CD209
Clone LWC06
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CIRE.
Gene ID (Entrez) 30835
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry