Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: eB-h209, Novus Biologicals™

$307.00 - $384.00

Specifications

Antigen DC-SIGN/CD209
Clone eB-h209
Immunogen The immunogen for this antibody was CD209.
Gene ID (Entrez) 30835
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14332701

 
novus biologicals canada
NBP14332701MG
0.1mg Each for $384.00

NBP14332703

 
novus biologicals canada
NBP1433270025MG
0.025mg Each for $307.00
Description & Specifications

Specifications

Antigen DC-SIGN/CD209
Clone eB-h209
Immunogen The immunogen for this antibody was CD209.
Gene ID (Entrez) 30835
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation