Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: eB-h209, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433270025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14332703

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunoprecipitation
Antigen DC-SIGN/CD209
Host Species Rat
Regulatory Status RUO
Formulation Phosphate buffer pH 7.2
Quantity 0.025mg
Cross Reactivity Human
Gene ID (Entrez) 30835
Monoclonal or Polyclonal Monoclonal
Immunogen The immunogen for this antibody was CD209.
Format Affinity Purified
Conjugate Unlabeled
Purification Method Affinity Purified
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Clone eB-h209
Isotype IgG2a, κ