Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: eB-h209, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14332701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14332701

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Primary or Secondary Primary
Immunogen The immunogen for this antibody was CD209.
Format Affinity Purified
Antigen DC-SIGN/CD209
Host Species Rat
Clone eB-h209
Isotype IgG2a, κ
Conjugate Unlabeled
Applications Flow Cytometry
Applications Immunoprecipitation
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 30835
Purification Method Affinity Purified
Quantity 0.1mg