Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: 5H10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433290025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14332903

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Antigen DC-SIGN/CD209
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 30835
Clone 5H10
Concentration 0.5mg/mL
Conjugate Unlabeled
Immunogen The immunogen for this antibody was CIRE.
Purification Method Affinity Purified
Regulatory Status RUO
Quantity 0.025mg