Please login to your online account to display your discounted pricing

Rat anti-DC-SIGN/CD209, Clone: 5H10, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14332901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14332901

  • / Each
 

Description & Specifications

Specifications

Applications Immunoprecipitation
Applications Western Blotting
Host Species Rat
Cross Reactivity Mouse
Format Affinity Purified
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Isotype IgG2a
Antigen DC-SIGN/CD209
Monoclonal or Polyclonal Monoclonal
Formulation Phosphate buffer pH 7.2
Concentration 0.5mg/mL
Quantity 0.1mg
Regulatory Status RUO
Clone 5H10
Purification Method Affinity Purified
Immunogen The immunogen for this antibody was CIRE.
Conjugate Unlabeled
Gene ID (Entrez) 30835