Please login to your online account to display your discounted pricing

Rat anti-Common gamma Chain/IL-2 R gamma, Clone: TUGh4, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen Common gamma Chain/IL-2 R gamma
Clone TUGh4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD132.
Gene ID (Entrez) 3561
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14358501

 
novus biologicals canada
NBP14358501MG
0.1mg Each for $252.00

NBP14358503

 
novus biologicals canada
NBP1435850025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen Common gamma Chain/IL-2 R gamma
Clone TUGh4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD132.
Gene ID (Entrez) 3561
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Isotype IgG2b, λ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry