Please login to your online account to display your discounted pricing

Rat anti-ChemR23/CMKLR1, Clone: BZ194, Novus Biologicals™

$274.00 - $450.00

Specifications

Antigen ChemR23/CMKLR1
Clone BZ194
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CMKLR1.
Gene ID (Entrez) 1240
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14323301

 
novus biologicals canada
NBP14323301MG
0.1mg Each for $329.00

NBP14323305

 
novus biologicals canada
NBP143233005MG
0.05mg Each for $274.00

NBP1432335

 
novus biologicals canada
NBP14323305MG
0.5mg Each for $450.00
Description & Specifications

Specifications

Antigen ChemR23/CMKLR1
Clone BZ194
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CMKLR1.
Gene ID (Entrez) 1240
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry