Please login to your online account to display your discounted pricing

Rat anti-ChemR23/CMKLR1, Clone: BZ194, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143233005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14323305

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Immunogen The immunogen for this antibody was CMKLR1.
Antigen ChemR23/CMKLR1
Formulation PBS [pH 7.2]
Host Species Rat
Applications Flow Cytometry
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1240
Clone BZ194
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse