Please login to your online account to display your discounted pricing

Rat anti-ChemR23/CMKLR1, Clone: BZ194, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14323301MG

 View all options for this product


For Research Use Only

Catalog No. NBP14323301


Description & Specifications


Antigen ChemR23/CMKLR1
Applications Flow Cytometry
Clone BZ194
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Rat
Immunogen The immunogen for this antibody was CMKLR1.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1240