Please login to your online account to display your discounted pricing

Rat anti-CD98, Clone: RL388, Novus Biologicals™

$214.00 - $278.00

Specifications

Antigen CD98
Clone RL388
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD98.
Gene ID (Entrez) 6520
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14337601

 
novus biologicals canada
NBP14337601MG
0.1mg Each for $278.00

NBP14337605

 
novus biologicals canada
NBP143376005MG
0.05mg Each for $214.00
Description & Specifications

Specifications

Antigen CD98
Clone RL388
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD98.
Gene ID (Entrez) 6520
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation