Please login to your online account to display your discounted pricing

Rat anti-CD98, Clone: RL388, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143376005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14337605

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 6520
Quantity 0.05mg
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.
Formulation Phosphate buffer pH 7.2
Isotype IgG2a, κ
Cross Reactivity Mouse
Antigen CD98
Immunogen The immunogen for this antibody was CD98.
Primary or Secondary Primary
Format Affinity Purified
Clone RL388
Concentration 0.5mg/mL
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunoprecipitation
Host Species Rat
Purification Method Affinity Purified