Please login to your online account to display your discounted pricing

Rat anti-CD98, Clone: RL388, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14398401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14398401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Formulation Phosphate buffer pH 7.2
Cross Reactivity Mouse
Format Affinity Purified
Antigen CD98
Host Species Rat
Isotype IgG2a, κ
Quantity 0.1mg
Storage Requirements Store at 4°C. Do not freeze.
Immunogen The immunogen for this antibody was CD98.
Gene ID (Entrez) 6520
Conjugate FITC
Regulatory Status RUO
Concentration 0.5mg/mL
Purification Method Affinity Purified
Clone RL388