Please login to your online account to display your discounted pricing

Rat anti-CD98, Clone: RL388, FITC, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14398405MG

 View all options for this product


For Research Use Only

Catalog No. NBP1439845


Description & Specifications


Antigen CD98
Applications Flow Cytometry
Clone RL388
Concentration 0.5mg/mL
Conjugate FITC
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was CD98.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6520