Please login to your online account to display your discounted pricing

Rat anti-CD98, Clone: RL388, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143984005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14398405

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Host Species Rat
Conjugate FITC
Formulation Phosphate buffer pH 7.2
Gene ID (Entrez) 6520
Concentration 0.5mg/mL
Primary or Secondary Primary
Clone RL388
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Format Affinity Purified
Immunogen The immunogen for this antibody was CD98.
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Antigen CD98
Quantity 0.05mg
Purification Method Affinity Purified
Cross Reactivity Mouse