Please login to your online account to display your discounted pricing

Rat anti-CD90.2, Clone: 53-2.1, Novus Biologicals™

$252.00 - $329.00

Specifications

Antigen CD90.2
Clone 53-2.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD90.2.
Gene ID (Entrez) 7070
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14338001

 
novus biologicals canada
NBP14338001MG
0.1mg Each for $285.00

NBP14338005

 
novus biologicals canada
NBP143380005MG
0.05mg Each for $252.00

NBP1433805

 
novus biologicals canada
NBP14338005MG
0.5mg Each for $329.00
Description & Specifications

Specifications

Antigen CD90.2
Clone 53-2.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD90.2.
Gene ID (Entrez) 7070
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation