Please login to your online account to display your discounted pricing

Rat anti-CD90.2, Clone: 53-2.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143380005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14338005

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7070
Host Species Rat
Storage Requirements Store at 4°C. Do not freeze.
Isotype IgG2a, κ
Regulatory Status RUO
Primary or Secondary Primary
Clone 53-2.1
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Cross Reactivity Mouse
Format Affinity Purified
Antigen CD90.2
Quantity 0.05mg
Formulation Phosphate buffer pH 7.2
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Immunogen The immunogen for this antibody was CD90.2.