Please login to your online account to display your discounted pricing

Rat anti-CD90.2, Clone: 53-2.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14338005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433805

  • / Each
 

Description & Specifications

Specifications

Antigen CD90.2
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen The immunogen for this antibody was CD90.2.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7070
Concentration 0.5mg/mL
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Clone 53-2.1
Conjugate Unlabeled