missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-CD90.2, Clone: 53-2.1, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD90.2
Clone 53-2.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD90.2.
Gene ID (Entrez) 7070
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14360901 Novus Biologicals Canada
NBP14360901MG
0.1mg Each for $131.00
NBP1436095 Novus Biologicals Canada
NBP14360905MG
0.5mg Each for $252.00
Specifications Description & Specifications

Specifications

CD90.2
0.5mg/mL
7070
Monoclonal
Store at 4°C. Do not freeze.
IgG2a, κ
Affinity Purified
Flow Cytometry
53-2.1
The immunogen for this antibody was CD90.2.
Phosphate buffer pH 7.2
Primary
Rat
Biotin
Affinity Purified