Please login to your online account to display your discounted pricing

Rat anti-CD90.2, Clone: 30-H12, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen CD90.2
Clone 30-H12
Concentration 0.5mg/mL
Immunogen Mouse CD90.2/Thy-1.2 alloantigen
Gene ID (Entrez) 7070
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14360801

 
novus biologicals canada
NBP14360801MG
0.1mg Each for $131.00

NBP1436085

 
novus biologicals canada
NBP14360805MG
0.5mg Each for $252.00
Description & Specifications

Specifications

Antigen CD90.2
Clone 30-H12
Concentration 0.5mg/mL
Immunogen Mouse CD90.2/Thy-1.2 alloantigen
Gene ID (Entrez) 7070
Formulation Phosphate buffer pH 7.2. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry