Please login to your online account to display your discounted pricing

Rat anti-CD9, Clone: KMC8, Novus Biologicals™

$219.00 - $406.00

Specifications

Antigen CD9
Clone KMC8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14344501

 
novus biologicals canada
NBP14344501MG
0.1mg Each for $285.00

NBP14344505

 
novus biologicals canada
NBP143445005MG
0.05mg Each for $219.00

NBP1434455

 
novus biologicals canada
NBP14344505MG
0.5mg Each for $406.00
Description & Specifications

Specifications

Antigen CD9
Clone KMC8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Western Blotting