Please login to your online account to display your discounted pricing

Rat anti-CD9, Clone: KMC8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14344501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344501

  • / Each
 

Description & Specifications

Specifications

Antigen CD9
Formulation PBS [pH 7.2]
Applications Flow Cytometry
Applications Western Blotting
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Immunogen The immunogen for this antibody was CD9.
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 928
Conjugate Unlabeled
Purification Method Affinity Purified
Clone KMC8
Concentration 0.5mg/mL