Please login to your online account to display your discounted pricing

Rat anti-CD9, Clone: KMC8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14344505MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1434455

  • / Each
 

Description & Specifications

Specifications

Antigen CD9
Applications Flow Cytometry
Applications Western Blotting
Quantity 0.5mg
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Rat
Isotype IgG2a, κ
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 928
Conjugate Unlabeled
Immunogen The immunogen for this antibody was CD9.
Storage Requirements Store at 4°C. Do not freeze.
Clone KMC8
Concentration 0.5mg/mL