Please login to your online account to display your discounted pricing

Rat anti-CD9, Clone: KMC8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143445005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344505

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Western Blotting
Antigen CD9
Host Species Rat
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Mouse
Formulation PBS [pH 7.2]
Immunogen The immunogen for this antibody was CD9.
Isotype IgG2a, κ
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone KMC8
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Gene ID (Entrez) 928