Please login to your online account to display your discounted pricing

Rat anti-CD9, Clone: KMC8, FITC, Novus Biologicals™

$175.00 - $197.00

Specifications

Antigen CD9
Clone KMC8
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14403501

 
novus biologicals canada
NBP14403501MG
0.1mg Each for $197.00

NBP14403505

 
novus biologicals canada
NBP144035005MG
0.05mg Each for $175.00
Description & Specifications

Specifications

Antigen CD9
Clone KMC8
Immunogen The immunogen for this antibody was CD9.
Gene ID (Entrez) 928
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry