Please login to your online account to display your discounted pricing

Rat anti-CD8 beta, Clone: H35-17.2, Novus Biologicals™

$219.00 - $285.00

Specifications

Antigen CD8 beta
Clone H35-17.2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD8.
Gene ID (Entrez) 926
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14344905

 
novus biologicals canada
NBP143449005MG
0.05mg Each for $219.00

NBP14344901

 
novus biologicals canada
NBP14344901MG
0.1mg Each for $285.00
Description & Specifications

Specifications

Antigen CD8 beta
Clone H35-17.2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD8.
Gene ID (Entrez) 926
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation