Please login to your online account to display your discounted pricing

Rat anti-CD8 beta, Clone: H35-17.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143449005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14344905

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Antigen CD8 beta
Immunogen The immunogen for this antibody was CD8.
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Concentration 0.5mg/mL
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Rat
Isotype IgG2b, κ
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Clone H35-17.2
Gene ID (Entrez) 926
Conjugate Unlabeled