Please login to your online account to display your discounted pricing

Rat anti-CD8 alpha (p Ser123, p Ser124, p ser123, p ser124), Clone: 53-6.7, PE-Cy5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200086100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20008610

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate PE-Cy5
Antigen CD8 alpha (p Ser123, p Ser124, p ser123, p ser124)
Format Affinity Purified
Host Species Rat
Immunogen The immunogen for this antibody is CD8 alpha.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 100μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 925
Clone 53-6.7
Concentration 0.2mg/mL
Cross Reactivity Mouse