Please login to your online account to display your discounted pricing

Rat anti-CD8 alpha (p Ser123, p Ser124, p ser123, p ser124), Clone: 53-6.7, Cy5, Novus Biologicals™

$321.00 - $418.00

Specifications

Antigen CD8 alpha (p Ser123, p Ser124, p ser123, p ser124)
Clone 53-6.7
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is CD8 alpha.
Gene ID (Entrez) 925
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20005126

 
novus biologicals canada
NBP200051200UG
200μg Each for $418.00

NBP20005110

 
novus biologicals canada
NBP200051100UG
100μg Each for $321.00
Description & Specifications

Specifications

Antigen CD8 alpha (p Ser123, p Ser124, p ser123, p ser124)
Clone 53-6.7
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is CD8 alpha.
Gene ID (Entrez) 925
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry