Please login to your online account to display your discounted pricing

Rat anti-CD8 alpha (p Ser123, p Ser124, p ser123, p ser124), Clone: 53-6.7, Cy5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200051200UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20005126

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Format Affinity Purified
Immunogen The immunogen for this antibody is CD8 alpha.
Host Species Rat
Regulatory Status RUO
Quantity 200μg
Clone 53-6.7
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Isotype IgG2a, κ
Concentration 0.2mg/mL
Purification Method Affinity Purified
Conjugate Cy5
Antigen CD8 alpha (p Ser123, p Ser124, p ser123, p ser124)
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 925