Please login to your online account to display your discounted pricing

Rat anti-CD8 alpha (p Ser123, p Ser124, p ser123, p ser124), Clone: 53-6.7, APC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200085200UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20008526

  • / Each
 

Description & Specifications

Specifications

Antigen CD8 alpha (p Ser123, p Ser124, p ser123, p ser124)
Applications Flow Cytometry
Immunogen The immunogen for this antibody is CD8 alpha.
Isotype IgG2a, κ
Quantity 200μg
Primary or Secondary Primary
Format Affinity Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 925
Concentration 0.2mg/mL
Conjugate APC
Cross Reactivity Mouse
Host Species Rat
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Clone 53-6.7