Please login to your online account to display your discounted pricing

Rat anti-CD8 alpha, Clone: 53-6.7, FITC, Novus Biologicals™

$171.00 - $386.00

Specifications

Antigen CD8 alpha
Clone 53-6.7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD8 alpha.
Gene ID (Entrez) 925
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20008910

 
novus biologicals canada
NBP200089100UG
100μg Each for $171.00

NBP20008955

 
novus biologicals canada
NBP200089500UG
500μg Each for $386.00
Description & Specifications

Specifications

Antigen CD8 alpha
Clone 53-6.7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD8 alpha.
Gene ID (Entrez) 925
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry