Please login to your online account to display your discounted pricing

Rat anti-CD8 alpha, Clone: 53-6.7, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200089100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20008910

  • / Each
 

Description & Specifications

Specifications

Quantity 100μg
Regulatory Status RUO
Host Species Rat
Format Affinity Purified
Antigen CD8 alpha
Isotype IgG2a, κ
Applications Flow Cytometry
Storage Requirements Store at 4°C in the dark.
Clone 53-6.7
Conjugate FITC
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD8 alpha.
Purification Method Affinity Purified
Primary or Secondary Primary
Gene ID (Entrez) 925
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse