Please login to your online account to display your discounted pricing

Rat anti-CD8 alpha, Clone: 53-6.7, FITC, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP200089500UG

 View all options for this product


For Research Use Only

Catalog No. NBP20008955


Description & Specifications


Antigen CD8 alpha
Applications Flow Cytometry
Clone 53-6.7
Concentration 0.5mg/mL
Conjugate FITC
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Immunogen The immunogen for this antibody is CD8 alpha.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 500μg
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 925