Please login to your online account to display your discounted pricing

Rat anti-CD63, Clone: 7G4.2E8, Novus Biologicals™

$109.00 - $307.00

Specifications

Antigen CD63
Clone 7G4.2E8
Concentration 1.0mg/mL
Immunogen Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731]
Gene Accession No. P41731
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236567

 
novus biologicals canada
NBP236567
0.1mg Each for $307.00

NBP236567SS

 
novus biologicals canada
NBP236567SS
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen CD63
Clone 7G4.2E8
Concentration 1.0mg/mL
Immunogen Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731]
Gene Accession No. P41731
Gene ID (Entrez) 967
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rat
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2a, λ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow (Intracellular)
Applications Flow Cytometry
Applications Western Blotting