Please login to your online account to display your discounted pricing

Rat anti-CD63, Clone: 7G4.2E8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP236567SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP236567SS

  • / Each
 

Description & Specifications

Specifications

Antigen CD63
Isotype IgG2a, λ
Host Species Rat
Clone 7G4.2E8
Applications Flow (Intracellular)
Applications Flow Cytometry
Applications Western Blotting
Gene Accession No. P41731
Cross Reactivity Human
Cross Reactivity Mouse
Conjugate Unlabeled
Format Purified
Formulation PBS. Protein G purified.
Concentration 1.0mg/mL
Quantity 0.025mg
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Immunogen Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731]
Purification Method Purified
Gene ID (Entrez) 967
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.