Please login to your online account to display your discounted pricing

Rat anti-CD45RB, Clone: C363.16A, Biotin, Novus Biologicals™

$128.00 - $321.00

Specifications

Antigen CD45RB
Clone C363.16A
Concentration 0.2mg/mL
Immunogen Cloned mouse Th2 cell lines (1)
Gene ID (Entrez) 5788
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14364001

 
novus biologicals canada
NBP14364001MG
0.1mg Each for $128.00

NBP14364005

 
novus biologicals canada
NBP143640005MG
0.05mg Each for $128.00

NBP1436405

 
novus biologicals canada
NBP14364005MG
0.5mg Each for $321.00
Description & Specifications

Specifications

Antigen CD45RB
Clone C363.16A
Concentration 0.2mg/mL
Immunogen Cloned mouse Th2 cell lines (1)
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry