Please login to your online account to display your discounted pricing

Rat anti-CD45RB, Clone: C363.16A, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14364001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14364001

  • / Each
 

Description & Specifications

Specifications

Antigen CD45RB
Applications Flow Cytometry
Clone C363.16A
Concentration 0.2mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Format Purified
Formulation Phosphate buffer pH 7.2. IgG purified.
Host Species Rat
Immunogen Cloned mouse Th2 cell lines (1)
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5788
Test Specificity An exon B-dependent epitope of CD45 glycoprotein