Please login to your online account to display your discounted pricing

Rat anti-CD45RB, Clone: C363.16A, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143640005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14364005

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5788
Conjugate Biotin
Antigen CD45RB
Format Purified
Formulation Phosphate buffer pH 7.2. IgG purified.
Host Species Rat
Immunogen Cloned mouse Th2 cell lines (1)
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity An exon B-dependent epitope of CD45 glycoprotein
Clone C363.16A
Concentration 0.2mg/mL
Cross Reactivity Mouse
Applications Flow Cytometry
Isotype IgG2a, κ
Purification Method Purified
Storage Requirements Store at 4°C. Do not freeze.