Please login to your online account to display your discounted pricing

Rat anti-CD45RB, Clone: C363.16A, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14364005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1436405

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Format Purified
Antigen CD45RB
Gene ID (Entrez) 5788
Concentration 0.2mg/mL
Immunogen Cloned mouse Th2 cell lines (1)
Monoclonal or Polyclonal Monoclonal
Formulation Phosphate buffer pH 7.2. IgG purified.
Quantity 0.5mg
Clone C363.16A
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Biotin
Regulatory Status RUO
Isotype IgG2a, κ
Test Specificity An exon B-dependent epitope of CD45 glycoprotein
Purification Method Purified
Primary or Secondary Primary
Host Species Rat