Please login to your online account to display your discounted pricing

Rat anti-CD45, Clone: 30-F11, FITC, Novus Biologicals™

$128.00 - $321.00

Specifications

Antigen CD45
Clone 30-F11
Concentration 0.5mg/mL
Immunogen Mouse thymus or spleen
Gene ID (Entrez) 5788
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20011710

 
novus biologicals canada
NBP200117100UG
100μg Each for $128.00

NBP20011755

 
novus biologicals canada
NBP200117500UG
500μg Each for $321.00
Description & Specifications

Specifications

Antigen CD45
Clone 30-F11
Concentration 0.5mg/mL
Immunogen Mouse thymus or spleen
Gene ID (Entrez) 5788
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry