Please login to your online account to display your discounted pricing

Rat anti-CD45, Clone: 30-F11, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200117500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20011755

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Isotype IgG2b, κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity CD45 (30-F11)
Conjugate FITC
Cross Reactivity Mouse
Antigen CD45
Format Affinity Purified
Host Species Rat
Immunogen Mouse thymus or spleen
Purification Method Affinity Purified
Quantity 500μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 5788
Clone 30-F11
Concentration 0.5mg/mL