Please login to your online account to display your discounted pricing

Rat anti-CD45, Clone: 30-F11, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200117100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20011710

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Conjugate FITC
Cross Reactivity Mouse
Regulatory Status RUO
Applications Flow Cytometry
Concentration 0.5mg/mL
Purification Method Affinity Purified
Isotype IgG2b, κ
Antigen CD45
Format Affinity Purified
Quantity 100μg
Gene ID (Entrez) 5788
Primary or Secondary Primary
Clone 30-F11
Host Species Rat
Immunogen Mouse thymus or spleen
Test Specificity CD45 (30-F11)
Storage Requirements Store at 4°C. Do not freeze.