Please login to your online account to display your discounted pricing

Rat anti-CD45, Clone: 30-F11, Biotin, Novus Biologicals™

$96.00 - $171.00

Specifications

Antigen CD45
Clone 30-F11
Immunogen Mouse thymus or spleen
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14364401

 
novus biologicals canada
NBP14364401MG
0.1mg Each for $171.00

NBP14364405

 
novus biologicals canada
NBP143644005MG
0.05mg Each for $96.00
Description & Specifications

Specifications

Antigen CD45
Clone 30-F11
Immunogen Mouse thymus or spleen
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry