Please login to your online account to display your discounted pricing

Rat anti-CD45, Clone: 30-F11, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143644005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14364405

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Applications Flow Cytometry
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Antigen CD45
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen Mouse thymus or spleen
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 5788
Test Specificity CD45 (30-F11)
Clone 30-F11
Isotype IgG2b, κ
Conjugate Biotin