Please login to your online account to display your discounted pricing

Rat anti-CD45, Clone: 30-F11, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14364401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14364401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Format Affinity Purified
Antigen CD45
Isotype IgG2b, κ
Immunogen Mouse thymus or spleen
Primary or Secondary Primary
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity CD45 (30-F11)
Cross Reactivity Mouse
Purification Method Affinity Purified
Formulation Phosphate buffer pH 7.2
Clone 30-F11
Gene ID (Entrez) 5788
Host Species Rat
Quantity 0.1mg
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Biotin