Please login to your online account to display your discounted pricing

Rat anti-CD44, Clone: IM7, Cy5, R-PE, Novus Biologicals™

$289.00 - $354.00

Specifications

Antigen CD44
Clone IM7
Concentration 1mg/mL
Immunogen Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M1
Gene ID (Entrez) 960
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP141266P5

 
novus biologicals canada
NBP141266PC550TESTS
50 Tests Each for $289.00

NBP141266PC

 
novus biologicals canada
NBP141266PC5100TESTS
100 Tests Each for $354.00
Description & Specifications

Specifications

Antigen CD44
Clone IM7
Concentration 1mg/mL
Immunogen Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M1
Gene ID (Entrez) 960
Formulation PBS [pH 7.4]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Rat
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2b
Conjugate Cy5, R-PE
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting