Please login to your online account to display your discounted pricing

Rat anti-CD44, Clone: IM7, Cy5, R-PE, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP141266PC550TESTS

 View all options for this product


For Research Use Only

Catalog No. NBP141266P5


Description & Specifications


Antigen CD44
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone IM7
Concentration 1mg/mL
Conjugate Cy5, R-PE
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS [pH 7.4]
Host Species Rat
Immunogen Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M1
Isotype IgG2b
Purification Method Purified
Quantity 50 Tests
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 960
Test Specificity The rat monoclonal antibody IM7 reacts with CD44 antigen (Phagocyte glycoprotein 1), an 80-95kDa transmembrane glycoprotein (hyaladherin family) present on the most of cells and tissues (leukocytes, endothelial cells, mesenchymal cells, etc.); it is negative on platelets and hepatocytes. The antibody reacts with all isoforms of mouse CD44.
Reconstitution Protein A purified